DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number ACR125312500
Price (MYR)
529.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
529.00
Add to cart
Chemical Identifiers
CAS771-97-1
IUPAC Namenaphthalene-2,3-diamine
Molecular FormulaC10H10N2
InChI KeyXTBLDMQMUSHDEN-UHFFFAOYSA-N
SMILESNC1=C(N)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Green-brown to brown
Melting point193°C to 199°C