DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number ACR125311000
Price (MYR)
381.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
381.00
Add to cart
Chemical Identifiers
CAS76116-20-6
IUPAC Name(1R)-1-(2-bromophenyl)ethan-1-ol
Molecular FormulaC8H9BrO
InChI KeyDZLZSFZSPIUINR-ZCFIWIBFSA-N
SMILESC[C@@H](O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%