Malonyl dichloride, 97%, Thermo Scientific Chemicals
Malonyl dichloride, 97%, Thermo Scientific Chemicals
Malonyl dichloride, 97%, Thermo Scientific Chemicals
Malonyl dichloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonyl dichloride, 97%, Thermo Scientific Chemicals
CAS: 1663-67-8 | C3H2Cl2O2 | 140.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR125280250
Price (MYR)
1,157.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,157.00
Add to cart
Chemical Identifiers
CAS51953-17-4
IUPAC Name4,5-dihydropyrimidin-4-one
Molecular FormulaC4H4N2O
InChI KeyTZYQTWHRLVDYPL-UHFFFAOYSA-N
SMILESO=C1CC=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)162.0-168.0?C