Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
CAS: 141-82-2 | C3H4O4 | 104.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR125262500
Price (MYR)
709.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
709.00
Add to cart
Chemical Identifiers
CAS868-14-4
IUPAC Namepotassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular FormulaC4H5KO6
InChI KeyKYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Identification (FTIR)Conforms
Optical Rotation32.3 ± 0.4? (C=10 in 1N NaOH)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%