Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
CAS: 141-82-2 | C3H4O4 | 104.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR125260010
Price (MYR)
1,578.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,578.00
Add to cart
Specifications
Chemical Name or MaterialMalonic acid
CAS141-82-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici