DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR125255000
Price (MYR)
457.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
457.00
Add to cart
Chemical Identifiers
CAS138-52-3
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
Molecular FormulaC13H18O7
InChI KeyNGFMICBWJRZIBI-UJPOAAIJSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=CC=C2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.5%
Optical Rotation-59.5? to -63.5? (c=5 in Water)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream