DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR125250010
Price (MYR)
548.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
548.00
Add to cart
Chemical Identifiers
CAS7758-87-4
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Barium (Ba)Passes test
Nitrate (NO3)Passes test
Sulfate (SO4)=<0.5 %
Arsenic (As)=<0.0003 %
Iron (Fe)=<0.04 %