Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR125231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2380-94-1
IUPAC Name1H-indol-4-ol
Molecular FormulaC8H7NO
InChI KeyNLMQHXUGJIAKTH-UHFFFAOYSA-N
SMILESOC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Green-grey to brown
Assay (HPLC)≥97.5%