Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR125230051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialMaleic acid
CAS110-16-7
Melting Point134.0°C to 138.0°C
ColorWhite
Health Hazard 2GHS H Statement: