L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number ACR125070025
Price (MYR)
892.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
892.00
Add to cart
Chemical Identifiers
CAS100-11-8
IUPAC Name1-(bromomethyl)-4-nitrobenzene
Molecular FormulaC7H6BrNO2
InChI KeyVOLRSQPSJGXRNJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystalline powder
Assay (GC)>97.0%
Solution Test5% in acetone: clear - no visible particlulates