DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number ACR125060250
Price (MYR)
385.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
385.00
Add to cart
Chemical Identifiers
CAS16523-54-9
IUPAC Namechlorodicyclohexylphosphane
Molecular FormulaC12H22ClP
InChI KeyAKJFBIZAEPTXIL-UHFFFAOYSA-N
SMILESClP(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric>=96.0 % (ex Cl)
Refractive index1.5310 to 1.5340 (20°C, 589 nm)