DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR125060025
Price (MYR)
1,402.00
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,402.00
Add to cart
Chemical Identifiers
CAS676-58-4
IUPAC Namechloro(methyl)magnesium
Molecular FormulaCH3ClMg
InChI KeyRQNMYNYHBQQZSP-UHFFFAOYSA-M
SMILESC[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay form precipitate on storage
Appearance (Color)Clear to faintly turbid
Concentration2.85-3.30M