DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR125060010
Price (MYR)
772.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
772.00
Add to cart
Chemical Identifiers
CAS619-42-1
IUPAC Namemethyl 4-bromobenzoate
Molecular FormulaC8H7BrO2
InChI KeyCZNGTXVOZOWWKM-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale gray
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)76.0-83.0?C