Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 5949-29-1 | C6H10O8 | 210.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR124912500
Price (MYR)
380.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS18497-13-7
IUPAC Nameplatinum(4+) dihydrogen hexahydrate hexachloride
Molecular FormulaCl6H14O6Pt
InChI KeyPIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay (special)Pt 37.5-39.9%
Total Metal Impurities≤0.05%
FormLump
Biological buffer for analysis
RUO – Research Use Only