Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid monohydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 5949-29-1 | C6H10O8 | 210.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR124910010
Price (MYR)
540.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
540.00
Add to cart
Chemical Identifiers
CAS562-90-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Biological buffer for analysis
RUO – Research Use Only