Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
CAS: 4445-07-2 | C24H42 | 330.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Glass bottle
Catalog number ACR124700500
Price (MYR)
6,719.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,719.00
Add to cart
Chemical Identifiers
CAS70380-73-3
IUPAC Name2-(1,3-oxazol-5-yl)pyridine
Molecular FormulaC8H6N2O
InChI KeyLUMKXMDGONYZHO-UHFFFAOYSA-N
SMILESO1C=NC=C1C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5955-1.6005 @ 20?C