Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Glass Bottle
Catalog number ACR124660011
Price (MYR)
662.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
662.00
Add to cart
Specifications
Chemical Name or MaterialNitric acid
ColorColorless
CAS7697-37-2
Melting Point-42.0°C
Boiling Point122.0°C
View more
For analysis
RUO – Research Use Only