β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR124532500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS56946-83-9
IUPAC Name2,5-dichlorothiophene-3-sulfonyl chloride
Molecular FormulaC4HCl3O2S2
InChI KeyJJKSHSHZJOWSEC-UHFFFAOYSA-N
SMILESClC1=CC(=C(Cl)S1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.5940-1.5990 @ 20?C