β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number ACR124530050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialβ-Nicotinamide adenine dinucleotide hydrate
CAS53-84-9
CAS Min %98.0
CAS Max %100.0
CAS53-84-9