β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Packaging:
Glass bottle
Catalog number ACR124530010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (special)Degree of Hydrolysis: 86.0-89.0%
Viscosity23.0-27.0 cP (4% solution)
Appearance (Color)White to off-white
Ash≤0.5% (Na₂O)