Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number ACR124280025
Price (MYR)
614.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
614.00
Add to cart
Chemical Identifiers
CAS100-42-5
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<20 APHA
Aldehyde=<0.02 % (as benzaldehyde)
View more
Volumetric standard solution
RUO – Research Use Only