Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid, Thermo Scientific Chemicals
CAS: 144-62-7 | C2H2O4 | 90.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number ACR124280010
Price (MYR)
418.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
418.00
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Volumetric standard solution
RUO – Research Use Only