Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR1242600101 LPlastic Bottle
Catalog number ACR124260010
Price (MYR)
432.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Chemical Identifiers
CAS2050-77-3
IUPAC Name1-iododecane
Molecular FormulaC10H21I
InChI KeySKIDNYUZJPMKFC-UHFFFAOYSA-N
SMILESCCCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.4830-1.4870 @ 20?C
Identification (FTIR)Conforms
StabilizerCopper Wire
View more
Volumetric standard solution
RUO – Research Use Only