Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number ACR124210010
Price (MYR)
457.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
457.00
Add to cart
Chemical Identifiers
CAS16466-61-8
IUPAC NameN'-[(tert-butoxy)carbonyl](tert-butoxy)carbohydrazide
Molecular FormulaC10H20N2O4
InChI KeyTYSZETYVESRFNT-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White
Assay (GC)≥98.0%
Volumetric standard solution
RUO – Research Use Only