Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Plastic Bottle
Catalog number ACR124190025
Price (MYR)
570.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
570.00
Add to cart
Chemical Identifiers
CAS4206-67-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4900-1.4930 @ 20?C
Volumetric standard solution
RUO – Research Use Only