Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
View all
Plastic bottle
Catalog number ACR124190010
Price (MYR)
427.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS9004-53-9
IUPAC Name(3R,4S,5S,6R)-2-{[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyFYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILESOC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](O)C(O[C@H]3[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Sulfated ash=<0.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Volumetric standard solution
RUO – Research Use Only