Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR124050025
Price (MYR)
554.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
554.00
Add to cart
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (on dried substance)
Residual solventsMeets the requirements
Arsenic (As)=<1 ppm
Infrared spectrumConforms
Loss on drying=<0.2 % (105°C, 2 h)