Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number ACR123990025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS10022-68-1
IUPAC Namecadmium(2+) bis(nitric acid)
Molecular FormulaCdH2N2O6
InChI KeyIZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Strength990 to 1010 µg/ml of Cd(2+) in 2 to 5% HNO3