Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl ether, 99+%, pure, stabilized with BHT, Thermo Scientific Chemicals
CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR123990010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS367-93-1
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular FormulaC9H18O5S
InChI KeyBPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILESCC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(5 % in water) Clear colorless
Appearance (Color)White
Infrared spectrumConforms
Specific optical rotation-29° to -33° (20°C, 589 nm) (c=1, water)
HPLC>=98.5 %