4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR123450050
Price (MYR)
378.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
378.00
Add to cart
Specifications
Chemical Name or Material4-Chlorophenylacetyl chloride
Name Note98%
CAS25026-34-0
CAS Min %95.0
ColorBrown to Yellow