Dicalite™ speed plus, Diatomaceous Filter-aid
Dicalite™ speed plus, Diatomaceous Filter-aid
Thermo Scientific Chemicals
Dicalite™ speed plus, Diatomaceous Filter-aid
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Catalog NumberQuantity
ACR1233800101 kg
Catalog number ACR123380010
Price (MYR)
445.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
445.00
Add to cart
Chemical Identifiers
CAS57-57-8
IUPAC Nameoxetan-2-one
Molecular FormulaC3H4O2
InChI KeyVEZXCJBBBCKRPI-UHFFFAOYSA-N
SMILESO=C1CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4105-1.4155 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless
Assay (GC)≥96.0% (non-U.S. specification)
View more
Filter aid
RUO – Research Use Only