Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic Acid, ca 48 wt. % Solution in Water (Tech.), Thermo Scientific Chemicals
CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number ACR123170010
Price (MYR)
677.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
677.00
Add to cart
Chemical Identifiers
CAS67-42-5
IUPAC Name3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular FormulaC14H24N2O10
InChI KeyDEFVIWRASFVYLL-UHFFFAOYSA-N
SMILESOC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration excess NaOH>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point240°C to 244°C
C-X bond formation reagent
RUO – Research Use Only