1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR123110050
Price (MYR)
72.58
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
72.58
Add to cart
Specifications
Chemical Name or Material1-(2-Chloroethyl)-4-methoxybenzene
Name Note97%
CAS18217-00-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning