Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2 kg
Packaging:
View all
Plastic bottle
Catalog number ACR122655000
Price (MYR)
459.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
459.00
Add to cart
Specifications
Chemical Name or MaterialIsophthalic acid
CAS121-91-5
Melting Point341.0°C to 343.0°C
ColorWhite
EINECS Number204-506-4