Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR122650020
Price (MYR)
953.00
Add to cart
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
953.00
Add to cart
Chemical Identifiers
CAS2967-54-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)198.0-208.0?C
Assay (GC)≥96.0%
Appearance (Color)White to pale cream