Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Thermo Scientific Chemicals
Isonipecotic Acid, 99%
CAS: 498-94-2 | C6H11NO2 | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR122620250
Price (MYR)
519.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
519.00
Add to cart
Chemical Identifiers
CAS873-73-4
IUPAC Name1-chloro-4-ethynylbenzene
Molecular FormulaC8H5Cl
InChI KeyLFZJRTMTKGYJRS-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Crystalline mass
Appearance (Color)Yellow to light brown
Infrared spectrumConforms
Melting point43°C to 47°C