Isatoic anhydride, 98%, Thermo Scientific Chemicals
Isatoic anhydride, 98%, Thermo Scientific Chemicals
Isatoic anhydride, 98%, Thermo Scientific Chemicals
Isatoic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isatoic anhydride, 98%, Thermo Scientific Chemicals
CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR122485000500 gPlastic bottle
Catalog number ACR122485000
Price (MYR)
898.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
898.00
Add to cart
Chemical Identifiers
CAS1079-66-9
IUPAC Namechlorodiphenylphosphane
Molecular FormulaC12H10ClP
InChI KeyXGRJZXREYAXTGV-UHFFFAOYSA-N
SMILESClP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay develop some turbidity or precipitate
Identification (FTIR)Conforms
Refractive Index1.6320-1.6380 @ 20?C
Appearance (Color)Colorless to yellow