2-Iodotoluene, 98%
2-Iodotoluene, 98%
2-Iodotoluene, 98%
Thermo Scientific Chemicals
2-Iodotoluene, 98%
CAS: 615-37-2 | C7H7I | 218.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR12243025025 mLGlass bottle
Catalog number ACR122430250
Price (MYR)
789.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
789.00
Add to cart
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Water=<0.5 % (K.F.)
Residue after evaporation=<0.005 %