Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number ACR122375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS768-95-6
IUPAC Nameadamantan-1-ol
Molecular FormulaC10H16O
InChI KeyVLLNJDMHDJRNFK-UHFFFAOYSA-N
SMILESOC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or needles
Alkylation reagent
RUO – Research Use Only