Iodoacetonitrile, 95%, Thermo Scientific Chemicals
Iodoacetonitrile, 95%, Thermo Scientific Chemicals
Iodoacetonitrile, 95%, Thermo Scientific Chemicals
Iodoacetonitrile, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoacetonitrile, 95%, Thermo Scientific Chemicals
CAS: 624-75-9 | C2H2IN | 166.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number ACR122290250
Price (MYR)
5,320.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,320.00
Add to cart
Specifications
Chemical Name or MaterialIodoacetonitrile
CAS624-75-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.