Iodoacetic acid, 97%, Thermo Scientific Chemicals
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Iodoacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoacetic acid, 97%, Thermo Scientific Chemicals
CAS: 64-69-7 | C2H3IO2 | 185.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR122280250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Residue after evaporation=<20 ppm