Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iminodibenzyl, 97%, Thermo Scientific Chemicals
CAS: 494-19-9 | C14H13N | 195.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR122071000
Price (MYR)
760.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
760.00
Add to cart
Specifications
Chemical Name or MaterialIminodibenzyl
Name Note97%
CAS494-19-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: