Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR122010250
Price (MYR)
787.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
787.00
Add to cart
Chemical Identifiers
CAS29968-78-3
IUPAC Name2-(4-nitrophenyl)ethan-1-aminium
Molecular FormulaC8H11N2O2
InChI KeyIOXOZOPLBFXYLM-UHFFFAOYSA-O
SMILES[NH3+]CCC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%