Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hypoxanthine, 99.5%, Thermo Scientific Chemicals
CAS: 68-94-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR122010050
Price (MYR)
472.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
472.00
Add to cart
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
UVat 220 nm A: =<0.20
UVat 230 nm A: =<0.10
UVat 240 nm A: =<0.04
UVat 250 nm A: =<0.01