Allopurinol, 98%
Allopurinol, 98%
Allopurinol, 98%
Thermo Scientific Chemicals
Allopurinol, 98%
CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR121830050
Price (MYR)
394.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Specifications
Chemical Name or MaterialAllopurinol
CAS315-30-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0