4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
CAS: 121-34-6 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR121351000
Price (MYR)
954.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
954.00
Add to cart
Chemical Identifiers
CAS6120-95-2
IUPAC Name1-phenylcyclopropane-1-carboxylic acid
Molecular FormulaC10H10O2
InChI KeyIWWCCNVRNHTGLV-UHFFFAOYSA-N
SMILESOC(=O)C1(CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)83.5-89.5?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%