N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals
N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-(2-Hydroxyethyl)piperazine, 98.5%, Thermo Scientific Chemicals
CAS: 103-76-4 | C6H14N2O | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR121191000
Price (MYR)
469.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS68988-92-1
IUPAC Name2,4,5,7-tetrabromo-9-(2-carboxyphenyl)-3,6-dihydroxy-10λ⁴-xanthen-10-ylium; 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium
Molecular FormulaC36H27Br4N3O5S
InChI KeyAXIKDPDWFVPGOD-UHFFFAOYSA-O
SMILESCN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C.OC(=O)C1=CC=CC=C1C1=C2C=C(Br)C(O)=C(Br)C2=[O+]C2=C(Br)C(O)=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
UV spectrumComplies
BSC CertificationPasses