Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
Actual product may vary
Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
Thermo Scientific Chemicals
Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers
These reusable Clear Glass Narrow Mouth Reagent Bottles with Glass Stoppers are strong, chemical resistant bottles with an enameled area for marking.
Have Questions?
Catalog NumberCapacity (Metric)
ACR121010250250 mL
Catalog number ACR121010250
Price (MYR)
504.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
504.00
EA
Add to cart
  • In conformity to DIN ISO 4796-2 Standard
  • Printed in white color
  • Sturdy ground neck
  • All sizes have a enameled area for marking
  • Strong and chemical resistant
  • Supplied with solid glass stopper
  • Autoclavable
Specifications
Capacity250 mL
Capacity (Metric)250 mL
Capacity (English)8.5 oz.
ColorTransparent
Diameter (English)2.8 ±0.1 in.
For Use With (Application)Storage
MaterialBorosilicate Glass (3.3)
Quantity12 Pk.
SterilitySterile
Certifications/ComplianceDIN ISO 4796-2
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)12.8 cm
Product TypeReagent Bottle
For Use With (Equipment)Chemicals
Height (English)5 in.
Chemical LabelCustom Write-on
Closure LinerUnlined
Closure MaterialBorosilicate Glass
Closure Size19/26
Diameter (Metric)70 ±2 mm
Closure TypeStopper
Unit SizeEA