4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
CAS: 5351-23-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR121001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS112-60-7
IUPAC Name2-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}ethan-1-ol
Molecular FormulaC8H18O5
InChI KeyUWHCKJMYHZGTIT-UHFFFAOYSA-N
SMILESOCCOCCOCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4581 to 1.4600 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %