Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number ACR120861000
Price (MYR)
460.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
460.00
Add to cart
Chemical Identifiers
CAS5431-44-7
IUPAC Namepyridine-2,6-dicarbaldehyde
Molecular FormulaC7H5NO2
InChI KeyPMWXGSWIOOVHEQ-UHFFFAOYSA-N
SMILESO=CC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point122°C to 126°C
GC>=96.0 %
Appearance (Color)White to tan
Appearance (Form)Powder